siRNA Details
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002568.3 | NP_002559.2 | 11 | 2068 | ||
XM_005250861.2 | XP_005250918.1 | 11 | 2979 | |||
GenBank mRNA | AK095194.1 | - | 11 | 1903 | ||
AK130523.1 | - | 11 | 2068 | |||
AK298990.1 | - | 12 | 1784 | |||
AK303120.1 | - | 12 | 1792 | |||
BC015958.2 | - | 12 | 1963 | 3940309 | ||
BC023217.1 | - | 11 | 2046 | |||
BC023520.2 | - | 11 | 2047 | 4123269 | ||
BC041863.1 | - | 11 | 2692 | |||
Y00345.1 | - | 11 | 2057 |
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016120.3 | NP_057204.2 | 4 | 10111 | ||
NM_183353.2 | NP_899196.1 | 5 | 10173 | |||
GenBank mRNA | AK127880.1 | - | 1 | 1960 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
