siRNA Details
Gene Symbol: EEF1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037663.1 | NP_001032752.1 | 4 | 450 | ||
NM_001959.3 | NP_001950.1 | 3 | 678 | |||
NM_021121.3 | NP_066944.1 | 4 | 404 | |||
GenBank mRNA | AK291910.1 | - | 3 | 376 | ||
AK310096.1 | - | 3 | 835 | |||
BC000211.1 | - | 4 | 370 | 3353094 | ||
BC004931.1 | - | 1 | 339 | 3611581 | ||
BC029316.1 | - | 3 | 354 | |||
BC067787.1 | - | 3 | 377 | 5299844 | ||
BM747256.1 | - | 4 | 419 | |||
BT007079.1 | - | 3 | 292 | |||
BU565415.1 | - | 410 | ||||
CR456825.1 | - | 3 | 292 | |||
S81522.1 | - | 4 | 437 | |||
X60489.1 | - | 3 | 527 | |||
X60656.1 | - | 4 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
