siRNA Details
Gene Symbol: EEF1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037663.1 | NP_001032752.1 | 7 | 704 | ||
NM_001959.3 | NP_001950.1 | 6 | 932 | |||
NM_021121.3 | NP_066944.1 | 7 | 658 | |||
GenBank mRNA | AK291910.1 | - | 6 | 630 | ||
BC000211.1 | - | 7 | 624 | 3353094 | ||
BC004931.1 | - | 1 | 593 | 3611581 | ||
BC029316.1 | - | 6 | 608 | |||
BC067787.1 | - | 6 | 631 | 5299844 | ||
BT007079.1 | - | 6 | 546 | |||
CR456825.1 | - | 6 | 546 | |||
X60489.1 | - | 6 | 781 | |||
X60656.1 | - | 7 | 641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
