siRNA Details
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200001.1 | NP_001186930.1 | 3 | 602 | ||
NM_024408.3 | NP_077719.2 | 3 | 602 | |||
XM_005270901.3 | XP_005270958.1 | 4 | 2916 | |||
XM_011541519.2 | XP_011539821.1 | 3 | 5710 | |||
XM_011541520.2 | XP_011539822.1 | 3 | 2620 | |||
XM_017001372.1 | XP_016856861.1 | 3 | 272 | |||
XM_017001373.1 | XP_016856862.1 | 3 | 251 | |||
GenBank mRNA | AF308601.1 | - | 3 | 317 | ||
AK307843.1 | - | 2 | 444 | |||
BC071562.1 | - | 3 | 551 | 4822955 | ||
GQ231534.1 | - | 3 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
