siRNA Details
Gene Symbol: FAM231D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC136705.1 | - | 1 | 113 |
Gene Symbol: FAM231A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282321.1 | NP_001269250.1 | 1 | 21 |
Gene Symbol: FAM231B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310155.1 | NP_001297084.1 | 1 | 363 | ||
XM_016999988.1 | XP_016855477.1 | 2 | 3072 |
Gene Symbol: FAM231C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310138.1 | NP_001297067.1 | 1 | 21 |
Gene Symbol: LOC100996721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005277507.4 | XP_005277564.1 | 1 | 1040 |
Gene Symbol: LOC102723383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003079.1 | XP_016858568.1 | 1 | 1748 |
Gene Symbol: LOC388692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123662.1 | - | 1 | 2196 | ||
BC141801.1 | - | 1 | 956 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
