siRNA Details
Gene Symbol: C1orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122961.1 | NP_001116433.1 | 4 | 563 | ||
XM_005270448.3 | XP_005270505.1 | 3 | 466 | |||
XM_006710342.3 | XP_006710405.1 | 4 | 909 | |||
XM_006710343.3 | XP_006710406.1 | 3 | 792 | |||
XM_011540647.2 | XP_011538949.1 | 4 | 940 | |||
GenBank mRNA | BC127905.1 | - | 2 | 254 | ||
BG196327.1 | - | 2 | 226 | |||
BG772797.1 | - | 4 | 564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
