siRNA Details
Gene Symbol: C1orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122961.1 | NP_001116433.1 | 2, 3 | 344 | ||
XM_006710342.3 | XP_006710405.1 | 2, 3 | 690 | |||
XM_011540647.2 | XP_011538949.1 | 2, 3 | 721 | |||
GenBank mRNA | BG772797.1 | - | 2, 3 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
