siRNA Details
Gene Symbol: DVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004421.2 | NP_004412.2 | 13 | 1453 | ||
XM_005244731.3 | XP_005244788.1 | 13 | 1558 | |||
XM_005244732.3 | XP_005244789.1 | 13 | 1558 | |||
XM_005244733.3 | XP_005244790.1 | 13 | 1482 | |||
GenBank mRNA | AB209210.1 | - | 8 | 1476 | ||
AF006011.1 | - | 13 | 1409 | |||
AK093189.1 | - | 6 | 693 | |||
AK095867.1 | - | 6 | 671 | |||
AK293286.1 | - | 13 | 1259 | |||
AK300063.1 | - | 5 | 455 | |||
BC017225.2 | - | 11 | 1178 | |||
BC050454.1 | - | 7 | 798 | 6201003 | ||
U46461.1 | - | 13 | 1550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
