siRNA Details
Gene Symbol: KRTAP4-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033059.3 | NP_149048.2 | 1 | 259 | ||
GenBank mRNA | AJ406944.1 | - | 1 | 265 | ||
BC126131.1 | - | 1 | 241 | 8991847 | ||
BC130562.1 | - | 1 | 241 | 40146593 |
Gene Symbol: KRTAP4-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030976.1 | NP_112238.1 | 1 | 340 | ||
GenBank mRNA | AJ406938.1 | - | 1 | 39 | ||
AJ406945.1 | - | 1 | 304 |
Gene Symbol: KRTAP4-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322457.1 | NP_001309386.1 | 1 | 260 |
Gene Symbol: KRTAP4-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146041.1 | NP_001139513.1 | 1 | 370 | ||
GenBank mRNA | AJ406941.1 | - | 1 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
