siRNA Details
Gene Symbol: KRTAP4-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146041.1 | NP_001139513.1 | 1 | 356 | ||
GenBank mRNA | AJ406941.1 | - | 1 | 301 |
Gene Symbol: KRTAP4-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033061.3 | NP_149050.3 | 1 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
