siRNA Details
Gene Symbol: ARID1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017519.2 | NP_059989.2 | 17 | 4473 | ||
NM_020732.3 | NP_065783.3 | 18 | 4512 | |||
XM_005267069.4 | XP_005267126.2 | 18 | 4707 | |||
XM_011535984.2 | XP_011534286.2 | 21 | 4918 | |||
XM_011535988.2 | XP_011534290.1 | 11 | 1861 | |||
XM_017011103.1 | XP_016866592.1 | 20 | 4819 | |||
XM_017011104.1 | XP_016866593.1 | 20 | 4788 | |||
XM_017011105.1 | XP_016866594.1 | 20 | 4759 | |||
XM_017011106.1 | XP_016866595.1 | 19 | 4629 | |||
XM_017011107.1 | XP_016866596.1 | 19 | 4609 | |||
XM_017011109.1 | XP_016866598.1 | 16 | 7145 | |||
GenBank mRNA | AB033061.1 | - | 12 | 2220 | ||
AF253515.1 | - | 17 | 3667 | |||
AF259792.1 | - | 13 | 2415 | |||
AF468300.1 | - | 18 | 3042 | |||
AF521671.1 | - | 17 | 4260 | |||
AK302356.1 | - | 1 | 68 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
