siRNA Details
Gene Symbol: ARID1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017519.2 | NP_059989.2 | 17 | 4599 | ||
NM_020732.3 | NP_065783.3 | 18 | 4638 | |||
XM_005267069.4 | XP_005267126.2 | 18 | 4833 | |||
XM_011535984.2 | XP_011534286.2 | 21 | 5044 | |||
XM_011535988.2 | XP_011534290.1 | 11 | 1987 | |||
XM_017011103.1 | XP_016866592.1 | 20 | 4945 | |||
XM_017011104.1 | XP_016866593.1 | 20 | 4914 | |||
XM_017011105.1 | XP_016866594.1 | 20 | 4885 | |||
XM_017011106.1 | XP_016866595.1 | 19 | 4755 | |||
XM_017011107.1 | XP_016866596.1 | 19 | 4735 | |||
XM_017011109.1 | XP_016866598.1 | 16 | 7271 | |||
GenBank mRNA | AB033061.1 | - | 12 | 2346 | ||
AF259792.1 | - | 13 | 2541 | |||
AF468300.1 | - | 18 | 3168 | |||
AF521671.1 | - | 17 | 4386 | |||
AK302356.1 | - | 1 | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
