siRNA Details
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 1, 2 | 185 | ||
NM_201400.3 | NP_958802.1 | 1, 2 | 185 | |||
NM_201598.3 | NP_963892.1 | 1, 2 | 185 | |||
XM_005255157.4 | XP_005255214.1 | 1, 2 | 158 | |||
XM_005255158.4 | XP_005255215.1 | 2, 3 | 231 | |||
XM_011522404.2 | XP_011520706.1 | 2, 3 | 231 | |||
GenBank mRNA | AL522535.3 | - | 1, 2 | 107 | ||
AY037162.1 | - | 1, 2 | 153 | |||
BC010084.2 | - | 1, 2 | 105 | 2822323 | ||
BG024981.1 | - | 1, 2 | 153 | |||
BQ921410.1 | - | 1, 2 | 168 | |||
CA495371.1 | - | 1, 2 | 106 | |||
HY017433.1 | - | 1, 2 | 185 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 1, 2 | 130 | ||
NM_018172.2 | NP_060642.2 | 1, 2 | 130 | |||
NM_152563.2 | NP_689776.1 | 1, 2 | 130 | |||
GenBank mRNA | AK001523.1 | - | 1, 2 | 104 | ||
AK130709.1 | - | 1, 2 | 127 | |||
AK293221.1 | - | 1, 2 | 104 | |||
AK311428.1 | - | 1, 2 | 104 | |||
AK311518.1 | - | 1, 2 | 127 | |||
AK311675.1 | - | 1, 2 | 131 | |||
AK315454.1 | - | 1, 2 | 104 | |||
BC032519.1 | - | 1, 2 | 130 | 5494088 | ||
BQ921333.1 | - | 1, 2 | 100 | |||
DC312827.1 | - | 1, 2 | 130 |
Gene Symbol: FAM86C2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024249.1 | 1, 2 | 127 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 1, 2 | 113 | |||
NR_130741.1 | 1, 2 | 113 | ||||
NR_130742.1 | 1, 2 | 113 | ||||
GenBank mRNA | AA421090.1 | - | 1, 2 | 76 | ||
CN271531.1 | - | 2, 3 | 153 | |||
CN481473.1 | - | 1, 2 | 106 | |||
HY065247.1 | - | 1, 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
