siRNA Details
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 6 | 1092 | ||
NM_201400.3 | NP_958802.1 | 8 | 1275 | |||
NM_201598.3 | NP_963892.1 | 7 | 1173 | |||
XM_005255157.4 | XP_005255214.1 | 7 | 1167 | |||
XM_005255158.4 | XP_005255215.1 | 9 | 1321 | |||
XM_011522404.2 | XP_011520706.1 | 8 | 1240 | |||
GenBank mRNA | AY037162.1 | - | 7 | 1142 | ||
BC010084.2 | - | 8 | 1195 | 2822323 | ||
BF314717.1 | - | 518 | ||||
BI491350.1 | - | 1 | 191 | |||
BQ215173.1 | - | 2 | 339 |
Gene Symbol: ALG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ002699.1 | - | 1 | 174 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 5 | 718 | ||
NM_018172.2 | NP_060642.2 | 5 | 739 | |||
NM_152563.2 | NP_689776.1 | 4 | 637 | |||
GenBank mRNA | AK001523.1 | - | 5 | 713 | ||
AK130709.1 | - | 4 | 634 | |||
AK293221.1 | - | 4 | 753 | |||
AK311428.1 | - | 5 | 695 | |||
AK311518.1 | - | 5 | 718 | |||
BC032519.1 | - | 4 | 637 | 5494088 | ||
BQ921333.1 | - | 5 | 688 |
Gene Symbol: FAM86C2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024249.1 | 4 | 634 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 4 | 1547 | |||
NR_130741.1 | 4 | 601 | ||||
NR_130742.1 | 4 | 551 | ||||
GenBank mRNA | BM718734.1 | - | 2 | 378 | ||
CN481473.1 | - | 3 | 460 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 841 | |||
GenBank mRNA | AK124448.1 | - | 3 | 610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
