siRNA Details
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 6 | 1117 | ||
NM_201400.3 | NP_958802.1 | 8 | 1300 | |||
NM_201598.3 | NP_963892.1 | 7 | 1198 | |||
XM_005255157.4 | XP_005255214.1 | 7 | 1192 | |||
XM_005255158.4 | XP_005255215.1 | 9 | 1346 | |||
XM_011522404.2 | XP_011520706.1 | 8 | 1265 | |||
GenBank mRNA | AY037162.1 | - | 7 | 1167 | ||
BC010084.2 | - | 8 | 1220 | 2822323 | ||
BF314717.1 | - | 543 | ||||
BI491350.1 | - | 1 | 216 | |||
BQ215173.1 | - | 2 | 364 |
Gene Symbol: ALG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ002699.1 | - | 1 | 199 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 5 | 743 | ||
NM_018172.2 | NP_060642.2 | 5 | 764 | |||
NM_152563.2 | NP_689776.1 | 4 | 662 | |||
GenBank mRNA | AK001523.1 | - | 5 | 738 | ||
AK130709.1 | - | 4 | 659 | |||
AK293221.1 | - | 4 | 778 | |||
AK311428.1 | - | 5 | 720 | |||
AK311518.1 | - | 5 | 743 | |||
BC032519.1 | - | 4 | 662 | 5494088 | ||
BQ921333.1 | - | 5 | 713 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 4 | 1572 | |||
NR_130741.1 | 4 | 626 | ||||
NR_130742.1 | 4 | 576 | ||||
GenBank mRNA | BM718734.1 | - | 2 | 403 | ||
CN481473.1 | - | 3 | 485 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 5 | 911 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 866 | |||
GenBank mRNA | AK124448.1 | - | 3 | 635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
