siRNA Details
Gene Symbol: HGH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016458.3 | NP_057542.2 | 3 | 793 | ||
GenBank mRNA | AK098226.1 | - | 4 | 487 | ||
BC003035.1 | - | 3 | 793 | 2822563 | ||
BC009915.2 | - | 3 | 793 | 2822563 | ||
BC015471.1 | - | 4 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
