siRNA Details
Gene Symbol: CDH26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021810.4 | NP_068582.2 | 2 | 253 | ||
NM_177980.2 | NP_817089.1 | 14 | 2356 | |||
XM_011528969.2 | XP_011527271.1 | 14 | 2362 | |||
XM_011528970.2 | XP_011527272.1 | 8 | 1378 | |||
XM_017027994.1 | XP_016883483.1 | 14 | 3025 | |||
XM_017027995.1 | XP_016883484.1 | 14 | 3026 | |||
XM_017027996.1 | XP_016883485.1 | 14 | 2927 | |||
XM_017027997.1 | XP_016883486.1 | 14 | 3026 | |||
XM_017027998.1 | XP_016883487.1 | 14 | 3023 | |||
XM_017027999.1 | XP_016883488.1 | 14 | 3023 | |||
XM_017028000.1 | XP_016883489.1 | 13 | 2195 | |||
XM_017028001.1 | XP_016883490.1 | 14 | 3023 | |||
XM_017028002.1 | XP_016883491.1 | 14 | 3024 | |||
GenBank mRNA | AF169690.1 | - | 6 | 820 | ||
AK055202.1 | - | 14 | 2345 | |||
AK074477.1 | - | 2 | 241 | |||
BC062570.1 | - | 2 | 253 | |||
BC136306.1 | - | 14 | 2160 | 9020293 | ||
BC143931.1 | - | 14 | 2160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
