siRNA Details
Gene Symbol: CDH26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021810.4 | NP_068582.2 | 5 | 484 | ||
NM_177980.2 | NP_817089.1 | 17 | 2587 | |||
XM_011528969.2 | XP_011527271.1 | 17 | 2593 | |||
XM_011528970.2 | XP_011527272.1 | 11 | 1609 | |||
XM_017027994.1 | XP_016883483.1 | 17 | 3256 | |||
XM_017027995.1 | XP_016883484.1 | 16 | 3188 | |||
XM_017027996.1 | XP_016883485.1 | 17 | 3158 | |||
XM_017027997.1 | XP_016883486.1 | 15 | 3134 | |||
XM_017027998.1 | XP_016883487.1 | 17 | 3254 | |||
XM_017027999.1 | XP_016883488.1 | 16 | 3185 | |||
XM_017028000.1 | XP_016883489.1 | 16 | 2426 | |||
XM_017028001.1 | XP_016883490.1 | 15 | 3131 | |||
XM_017028002.1 | XP_016883491.1 | 17 | 3255 | |||
GenBank mRNA | AF169690.1 | - | 7 | 928 | ||
AK055202.1 | - | 17 | 2576 | |||
AK074477.1 | - | 5 | 472 | |||
BC062570.1 | - | 3 | 361 | |||
BC136306.1 | - | 17 | 2570 | 9020293 | ||
BC143931.1 | - | 17 | 2570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
