siRNA Details
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 6 | 836 | ||
NM_201400.3 | NP_958802.1 | 8 | 1019 | |||
NM_201598.3 | NP_963892.1 | 7 | 917 | |||
XM_005255157.4 | XP_005255214.1 | 7 | 911 | |||
XM_005255158.4 | XP_005255215.1 | 9 | 1065 | |||
XM_011522404.2 | XP_011520706.1 | 8 | 984 | |||
GenBank mRNA | AL522535.3 | - | 6 | 758 | ||
AY037162.1 | - | 7 | 885 | |||
BC010084.2 | - | 8 | 939 | 2822323 | ||
BF314717.1 | - | 262 | ||||
BQ215173.1 | - | 2 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
