siRNA Details
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 3 | 1048 | |||
GenBank mRNA | AK056232.1 | - | 3 | 889 | ||
AK057951.1 | - | 2 | 244 | |||
AK095183.1 | - | 3 | 242 | |||
AK290087.1 | - | 2 | 247 | |||
AK291582.1 | - | 3 | 257 | |||
AK295256.1 | - | 3 | 254 | |||
AK300161.1 | - | 4 | 470 | |||
BC048328.1 | - | 2 | 244 | |||
BC063555.1 | - | 3 | 446 | |||
BC110996.1 | - | 3 | 746 | |||
BM839085.1 | - | 2 | 243 | |||
BQ722707.1 | - | 2 | 258 | |||
BX452679.2 | - | 381 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 3 | 370 | |||
GenBank mRNA | CN404396.1 | - | 2 | 234 | ||
DA015248.1 | - | 3 | 256 | |||
DC319181.1 | - | 3 | 373 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 3 | 370 | ||
XM_011517658.2 | XP_011515960.1 | 3 | 487 | |||
XM_011517659.2 | XP_011515961.1 | 3 | 269 | |||
XM_011517660.2 | XP_011515962.1 | 3 | 266 | |||
XM_011517662.2 | XP_011515964.1 | 3 | 1773 | |||
XM_011517663.2 | XP_011515965.1 | 2 | 1622 | |||
XM_011517664.2 | XP_011515966.1 | 2 | 1619 | |||
XM_011517665.2 | XP_011515967.1 | 2 | 168 | |||
XM_011517671.2 | XP_011515973.2 | 3 | 487 | |||
XM_017014169.1 | XP_016869658.1 | 3 | 386 | |||
XM_017014170.1 | XP_016869659.1 | 3 | 449 | |||
XM_017014173.1 | XP_016869662.1 | 3 | 487 | |||
GenBank mRNA | AK024481.1 | - | 1 | 432 | ||
AK289708.1 | - | 3 | 373 | |||
AK309568.1 | - | 2 | 102 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 3 | 495 | |||
GenBank mRNA | AK292759.1 | - | 2 | 89 | ||
AK308574.1 | - | 2 | 104 | |||
AL137714.1 | - | 3 | 382 | |||
AY217347.1 | - | 2 | 927 | |||
BC082240.1 | - | 1 | 952 | |||
BX377099.2 | - | 3 | 260 | |||
BX537637.1 | - | 3 | 496 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 3 | 269 | |||
GenBank mRNA | AK294377.1 | - | 2 | 165 | ||
AK296182.1 | - | 4 | 405 | |||
AK302089.1 | - | 3 | 314 | |||
AK308540.1 | - | 5 | 524 | |||
AK309926.1 | - | 3 | 256 | |||
AK310121.1 | - | 3 | 243 | |||
AK310139.1 | - | 4 | 297 | |||
AK316487.1 | - | 2 | 458 | |||
BC094698.1 | - | 1 | 807 | |||
DA054875.1 | - | 3 | 277 | |||
DA143319.1 | - | 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
