siRNA Details
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 3, 4 | 521 | |||
GenBank mRNA | AK292759.1 | - | 2, 3 | 115 | ||
AK308574.1 | - | 2, 3 | 130 | |||
AL137714.1 | - | 3, 4 | 408 | |||
BC082240.1 | - | 1, 2 | 978 | |||
BX377099.2 | - | 3, 4 | 286 | |||
BX537637.1 | - | 3, 4 | 522 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 3, 4 | 396 | |||
GenBank mRNA | CN404396.1 | - | 2, 3 | 260 | ||
DA015248.1 | - | 3, 4 | 282 | |||
DC319181.1 | - | 3, 4 | 399 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 3, 4 | 396 | ||
XM_011517658.2 | XP_011515960.1 | 3, 4 | 513 | |||
XM_011517659.2 | XP_011515961.1 | 3, 4 | 295 | |||
XM_011517660.2 | XP_011515962.1 | 3, 4 | 292 | |||
XM_011517662.2 | XP_011515964.1 | 3, 4 | 1799 | |||
XM_011517663.2 | XP_011515965.1 | 2, 3 | 1648 | |||
XM_011517664.2 | XP_011515966.1 | 2, 3 | 1645 | |||
XM_011517665.2 | XP_011515967.1 | 2, 3 | 194 | |||
XM_011517671.2 | XP_011515973.2 | 3, 4 | 513 | |||
XM_017014169.1 | XP_016869658.1 | 3, 4 | 412 | |||
XM_017014170.1 | XP_016869659.1 | 3, 4 | 475 | |||
XM_017014173.1 | XP_016869662.1 | 3, 4 | 513 | |||
GenBank mRNA | AK024481.1 | - | 1 | 458 | ||
AK289708.1 | - | 3, 4 | 399 | |||
AK309568.1 | - | 2, 3 | 128 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 3, 4 | 1074 | |||
GenBank mRNA | AK056232.1 | - | 3, 4 | 915 | ||
AK057951.1 | - | 2, 3 | 270 | |||
AK095183.1 | - | 3, 4 | 268 | |||
AK290087.1 | - | 2, 3 | 273 | |||
AK291582.1 | - | 3, 4 | 283 | |||
AK295256.1 | - | 3, 4 | 280 | |||
AK300161.1 | - | 4, 5 | 496 | |||
BC048328.1 | - | 2, 3 | 270 | |||
BC063555.1 | - | 3, 4 | 472 | |||
BC110996.1 | - | 3, 4 | 772 | |||
BM839085.1 | - | 2, 3 | 269 | |||
BQ722707.1 | - | 2, 3 | 284 | |||
BX452679.2 | - | 407 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 3, 4 | 295 | |||
GenBank mRNA | AK294377.1 | - | 2, 3 | 191 | ||
AK296182.1 | - | 4, 5 | 431 | |||
AK302089.1 | - | 3, 4 | 340 | |||
AK308540.1 | - | 5, 6 | 550 | |||
AK309926.1 | - | 3, 4 | 282 | |||
AK310121.1 | - | 3 | 269 | |||
AK310139.1 | - | 4, 5 | 323 | |||
AK316487.1 | - | 2, 3 | 484 | |||
BC094698.1 | - | 1, 2 | 833 | |||
DA054875.1 | - | 3, 4 | 303 | |||
DA143319.1 | - | 3, 4 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
