siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 5 | 645 | ||
XM_011517658.2 | XP_011515960.1 | 5 | 762 | |||
XM_011517659.2 | XP_011515961.1 | 5 | 544 | |||
XM_011517660.2 | XP_011515962.1 | 5 | 541 | |||
XM_011517662.2 | XP_011515964.1 | 5 | 2048 | |||
XM_011517663.2 | XP_011515965.1 | 4 | 1897 | |||
XM_011517664.2 | XP_011515966.1 | 4 | 1894 | |||
XM_011517665.2 | XP_011515967.1 | 4 | 443 | |||
XM_011517666.2 | XP_011515968.1 | 4 | 442 | |||
XM_011517671.2 | XP_011515973.2 | 5 | 762 | |||
XM_017014169.1 | XP_016869658.1 | 5 | 661 | |||
XM_017014170.1 | XP_016869659.1 | 5 | 724 | |||
XM_017014171.1 | XP_016869660.1 | 4 | 472 | |||
XM_017014172.1 | XP_016869661.1 | 4 | 2050 | |||
XM_017014173.1 | XP_016869662.1 | 5 | 762 | |||
GenBank mRNA | AK024481.1 | - | 1 | 1085 | ||
AK289708.1 | - | 5 | 648 | |||
AK309568.1 | - | 4 | 377 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 5 | 645 | |||
GenBank mRNA | CN404396.1 | - | 4 | 509 | ||
DA015248.1 | - | 5 | 531 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 5 | 770 | |||
GenBank mRNA | AK292759.1 | - | 4 | 373 | ||
AK308574.1 | - | 4 | 379 | |||
AL137714.1 | - | 5 | 657 | |||
AY217347.1 | - | 4 | 1202 | |||
BC082240.1 | - | 3 | 1227 | |||
BC106020.1 | - | 2 | 194 | |||
BX377099.2 | - | 5 | 533 | |||
BX537637.1 | - | 5 | 771 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 5 | 1319 | |||
GenBank mRNA | AK056232.1 | - | 5 | 1164 | ||
AK057951.1 | - | 7 | 769 | |||
AK095183.1 | - | 5 | 526 | |||
AK291582.1 | - | 5 | 541 | |||
AK295256.1 | - | 5 | 538 | |||
AK300161.1 | - | 6 | 741 | |||
BC048328.1 | - | 4 | 519 | |||
BC063555.1 | - | 5 | 721 | |||
BC110996.1 | - | 5 | 1021 | |||
BQ722707.1 | - | 4 | 529 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 5 | 544 | |||
GenBank mRNA | AK092583.1 | - | 4 | 419 | ||
AK294377.1 | - | 4 | 440 | |||
AK296182.1 | - | 6 | 680 | |||
AK302089.1 | - | 5 | 598 | |||
AK309926.1 | - | 5 | 531 | |||
AK310121.1 | - | 5 | 418 | |||
AK310139.1 | - | 6 | 572 | |||
AK316487.1 | - | 4 | 733 | |||
BC063470.1 | - | 3 | 593 | |||
BC094698.1 | - | 3 | 1082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
