siRNA Details
Gene Symbol: LCE1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276331.1 | NP_001263260.1 | 3 | 186 | ||
NM_178351.3 | NP_848128.1 | 2 | 276 |
Gene Symbol: LCE1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178348.2 | NP_848125.1 | 1 | 201 |
Gene Symbol: LCE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178349.1 | NP_848126.1 | 1 | 701 | ||
GenBank mRNA | BC031811.1 | - | 1 | 701 | ||
BC104232.1 | - | 1 | 309 | 40032247 | ||
BC104233.1 | - | 1 | 311 | 40032251 | ||
BC133657.1 | - | 1 | 311 |
Gene Symbol: LCE1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178353.1 | NP_848130.1 | 2 | 278 | ||
GenBank mRNA | BC038391.1 | - | 2 | 278 |
Gene Symbol: LCE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178356.3 | NP_848133.1 | 1 | 203 | ||
GenBank mRNA | BC113446.1 | - | 1 | 203 | 8322498 | |
BC113448.1 | - | 1 | 203 | 8322500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
