siRNA Details
Gene Symbol: PDPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522521.2 | XP_011520823.1 | 1 | 2862 | ||
XM_011522523.2 | XP_011520825.1 | 1 | 2867 | |||
XM_017023271.1 | XP_016878760.1 | 1 | 2867 | |||
XM_017023275.1 | XP_016878764.1 | 1 | 2873 |
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL547558.3 | - | 2 | 168 | ||
AU122729.1 | - | 2 | 156 | |||
BX382279.2 | - | 2 | 156 | |||
BX477204.1 | - | 2 | 194 |
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273106.2 | XP_005273163.1 | 1 | 52 |
Gene Symbol: LOC100132272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124207.1 | - | 1 | 1536 |
Gene Symbol: MTDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178812.3 | NP_848927.2 | 12 | 7117 | ||
GenBank mRNA | AK123083.1 | - | 1 | 1612 |
Gene Symbol: ZNF785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152458.6 | NP_689671.2 | 3 | 2971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
