siRNA Details
Gene Symbol: SLC27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198580.2 | NP_940982.1 | 12 | 2378 | ||
XM_011528000.1 | XP_011526302.1 | 14 | 2555 | |||
XM_011528001.2 | XP_011526303.1 | 13 | 2354 | |||
XM_011528002.2 | XP_011526304.1 | 14 | 2335 | |||
XM_011528003.2 | XP_011526305.1 | 13 | 2548 | |||
XM_017026781.1 | XP_016882270.1 | 14 | 2492 | |||
GenBank mRNA | AK131101.1 | - | 6 | 3123 | ||
AK299852.1 | - | 13 | 2647 | |||
BC013149.1 | - | 6 | 809 | |||
BC059399.1 | - | 12 | 2331 | 30341529 |
Gene Symbol: LOC100128108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720812.3 | XP_006720875.1 | 1 | 513 |
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130807.2 | NP_570719.1 | 5 | 2230 | ||
XM_011527683.2 | XP_011525985.1 | 5 | 2307 | |||
GenBank mRNA | AK094732.1 | - | 2 | 1331 | ||
AL832474.1 | - | 5 | 2266 | |||
BC015049.1 | - | 4 | 2119 | 3919665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
