siRNA Details
Gene Symbol: GUCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284251.1 | NP_001271180.1 | 6 | 2469 | ||
NM_001284252.1 | NP_001271181.1 | 6 | 2466 | |||
NM_001284253.1 | NP_001271182.1 | 6 | 2337 | |||
NM_001284254.1 | NP_001271183.1 | 6 | 2504 | |||
NM_001284255.1 | NP_001271184.1 | 6 | 2375 | |||
NM_001284256.1 | NP_001271185.1 | 5 | 2265 | |||
NM_001284257.1 | NP_001271186.1 | 5 | 2262 | |||
NM_031444.3 | NP_113632.2 | 6 | 2507 | |||
NR_104286.1 | 4 | 2142 | ||||
XM_005261761.1 | XP_005261818.1 | 5 | 2225 | |||
GenBank mRNA | AK024723.1 | - | 1 | 219 | ||
AK025242.1 | - | 1 | 1369 | |||
BC002924.2 | - | 5 | 2162 | |||
BC053529.1 | - | 6 | 2161 | |||
BC070109.1 | - | 6 | 2510 | 4838028 |
Gene Symbol: A1BG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056201.1 | - | 2 | 682 | ||
EU794585.1 | - | 8 | 1934 |
Gene Symbol: AIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286588.1 | NP_001273517.1 | 4 | 2420 | ||
NM_001286589.1 | NP_001273518.1 | 4 | 2179 | |||
GenBank mRNA | BX640703.1 | - | 4 | 2121 |
Gene Symbol: ALDH7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001182.4 | NP_001173.2 | 18 | 4339 | ||
NM_001201377.1 | NP_001188306.1 | 18 | 4339 | |||
NM_001202404.1 | NP_001189333.2 | 16 | 4147 | |||
GenBank mRNA | AK123774.1 | - | 1 | 697 |
Gene Symbol: C8orf34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038877.1 | 3 | 426 |
Gene Symbol: CAMK2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308853.1 | - | 14 | 1493 |
Gene Symbol: CDK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ773564.1 | - | 1 | 143 |
Gene Symbol: CLCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048210.2 | NP_001041675.1 | 12 | 3811 | ||
NM_001278202.1 | NP_001265131.1 | 9 | 3319 | |||
NM_001278203.1 | NP_001265132.1 | 7 | 3127 | |||
NM_015127.4 | NP_055942.1 | 12 | 3671 | |||
GenBank mRNA | AB018304.2 | - | 11 | 3672 | ||
AK090563.1 | - | 8 | 1874 |
Gene Symbol: CNDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ183399.1 | - | 1 | 575 |
Gene Symbol: CREB3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK304259.1 | - | 3 | 1026 |
Gene Symbol: CRLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015986.3 | NP_057070.3 | 8 | 2099 | ||
NR_073118.1 | 7 | 1891 | ||||
XM_011524879.1 | XP_011523181.1 | 8 | 2273 | |||
GenBank mRNA | AF046059.1 | - | 8 | 2012 | ||
AF120151.1 | - | 7 | 2465 | |||
BC023567.2 | - | 8 | 2021 | 4309046 |
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028911.1 | - | 7 | 1002 |
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023777.1 | XP_016879266.1 | 1 | 266 |
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126264.1 | - | 14 | 2059 |
Gene Symbol: GLRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199868.1 | NP_001186797.1 | 12 | 1170 | ||
XM_017015474.1 | XP_016870963.1 | 13 | 1225 | |||
GenBank mRNA | AJ010841.1 | - | 12 | 1128 | ||
AK022131.1 | - | 12 | 1144 | |||
BI256922.1 | - | 7 | 511 |
Gene Symbol: LOC100131262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092421.1 | - | 1 | 1035 |
Gene Symbol: LOC105369662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094655.1 | - | 1 | 1108 |
Gene Symbol: LOC105375240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC036546.1 | - | 1 | 2499 |
Gene Symbol: LOC283575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092322.1 | - | 3 | 495 |
Gene Symbol: LOC285500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094945.1 | - | 2 | 558 |
Gene Symbol: LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094081.1 | - | 1 | 1160 |
Gene Symbol: MALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092004.1 | - | 1 | 1023 |
Gene Symbol: MTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123226.1 | NP_001116698.1 | 8 | 1449 | ||
GenBank mRNA | AK225828.1 | - | 8 | 1433 |
Gene Symbol: MVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF217536.1 | - | 3 | 299 |
Gene Symbol: OPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU617706.1 | - | 1 | 340 |
Gene Symbol: PECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG939782.1 | - | 1 | 328 |
Gene Symbol: PMM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209659.1 | - | 1 | 1997 |
Gene Symbol: RAD54L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322253.1 | NP_001309182.1 | 24 | 8654 | ||
NM_001322256.1 | NP_001309185.1 | 22 | 8544 | |||
NM_001322257.1 | NP_001309186.1 | 22 | 8446 | |||
NM_015106.3 | NP_055921.2 | 23 | 8606 | |||
XM_006713062.3 | XP_006713125.1 | 22 | 8353 | |||
XM_017005987.1 | XP_016861476.1 | 22 | 8382 | |||
XM_017005988.1 | XP_016861477.1 | 23 | 9942 |
Gene Symbol: RGS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008530.1 | XP_016864019.1 | 4 | 4140 |
Gene Symbol: SPTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209757.1 | - | 8 | 1172 |
Gene Symbol: SUCLA2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029610.1 | - | 2 | 493 |
Gene Symbol: SYCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097443.1 | - | 1 | 161 |
Gene Symbol: SYNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833542.1 | - | 1 | 1834 |
Gene Symbol: TAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716100.2 | XP_006716163.1 | 2 | 238 | ||
XM_011516542.2 | XP_011514844.1 | 2 | 247 |
Gene Symbol: TNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209338.1 | - | 2 | 1615 |
Gene Symbol: TRIM52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209243.1 | - | 1 | 2837 | ||
BC034154.1 | - | 1 | 271 |
Gene Symbol: TRIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK304759.1 | - | 4 | 633 |
Gene Symbol: TSPAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005981.3 | NP_005972.1 | 6 | 1348 | ||
XM_005269074.3 | XP_005269131.2 | 6 | 2273 | |||
XM_017019793.1 | XP_016875282.1 | 4 | 1077 | |||
GenBank mRNA | AK090502.1 | - | 4 | 1751 | ||
BC010377.1 | - | 6 | 1300 | 4294627 | ||
BX647402.1 | - | 4 | 1074 | |||
U01160.1 | - | 6 | 1296 |
Gene Symbol: TXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123361.1 | - | 1 | 454 |
Gene Symbol: UMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000373.3 | NP_000364.1 | 6 | 2117 | ||
NR_033434.1 | 5 | 1963 | ||||
NR_033437.1 | 7 | 2216 | ||||
GenBank mRNA | EU921886.1 | - | 6 | 2031 | ||
EU921887.1 | - | 5 | 1877 | |||
EU921889.1 | - | 6 | 1976 | |||
EU921890.1 | - | 6 | 2234 | |||
EU921891.1 | - | 7 | 2130 | |||
EU921892.1 | - | 6 | 1713 | |||
EU921893.1 | - | 6 | 1684 | |||
EU921895.1 | - | 3 | 1586 | |||
J03626.1 | - | 6 | 2115 |
Gene Symbol: VEZT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309943.1 | - | 10 | 2021 |
Gene Symbol: WIPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124974.1 | - | 1 | 516 |
Gene Symbol: ZNF252P-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026974.1 | 1 | 890 | |||
GenBank mRNA | AK092777.1 | - | 1 | 715 | ||
BC046439.1 | - | 1 | 890 | |||
BC122544.1 | - | 1 | 864 |
Gene Symbol: ZNF266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB095928.1 | - | 1 | 82 |
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 3971 | ||
NR_036599.1 | 5 | 4089 | ||||
NR_036600.1 | 3 | 3844 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 3946 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 4001 | |||
GenBank mRNA | AK091553.1 | - | 1 | 1876 | ||
AK092385.1 | - | 1 | 2778 | |||
AK131368.1 | - | 2 | 4566 | |||
AK131574.1 | - | 1 | 3356 | |||
AK304704.1 | - | 4 | 3933 | |||
BC140720.1 | - | 3 | 3870 | 9021588 | ||
BX640767.1 | - | 1 | 2999 | |||
CR933598.1 | - | 4 | 3931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
