siRNA Details
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000903.2 | NP_000894.1 | 6 | 1880 | ||
NM_001025433.1 | NP_001020604.1 | 5 | 1778 | |||
NM_001025434.1 | NP_001020605.1 | 5 | 1766 | |||
NM_001286137.1 | NP_001273066.1 | 4 | 1702 | |||
GenBank mRNA | BM787983.1 | - | 1 | 512 | ||
BM828301.1 | - | 1 | 109 | |||
J03934.1 | - | 6 | 1738 |
Gene Symbol: HPN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024561.1 | 4 | 3454 |
Gene Symbol: STX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY203924.1 | - | 1 | 260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
