siRNA Details
Gene Symbol: ZNF658B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003528.3 | 5 | 898 | |||
GenBank mRNA | AK095694.1 | - | 1 | 245 | ||
BC137089.1 | - | 1 | 245 | 9021086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
