siRNA Details
Gene Symbol: ERGIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016570.2 | NP_057654.2 | 8 | 688 | ||
GenBank mRNA | AF183410.1 | - | 8 | 639 | ||
AF216751.1 | - | 8 | 643 | |||
AF302767.1 | - | 8 | 641 | |||
AK074520.1 | - | 8 | 663 | |||
AK222799.1 | - | 8 | 628 | |||
AL834128.2 | - | 8 | 629 | |||
BC000887.1 | - | 8 | 641 | 3462408 | ||
BC107794.1 | - | 8 | 645 | 30319807 |
