siRNA Details
Gene Symbol: AP1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039569.1 | NP_001034658.1 | 5 | 3026 | ||
NR_110905.1 | 6 | 3065 | ||||
NR_110906.1 | 4 | 2917 | ||||
XM_011510600.2 | XP_011508902.1 | 4 | 2868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
