siRNA Details
Gene Symbol: MESDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB365572.1 | - | 1 | 411 |
Gene Symbol: SMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003652.3 | NP_001003652.1 | 11 | 6210 | ||
NM_001135937.2 | NP_001129409.1 | 10 | 6120 | |||
NM_005901.5 | NP_005892.1 | 11 | 6102 | |||
XM_005258259.3 | XP_005258316.1 | 12 | 6343 | |||
XM_006722451.3 | XP_006722514.1 | 12 | 6355 | |||
XM_011525984.2 | XP_011524286.1 | 11 | 5699 | |||
XM_011525985.2 | XP_011524287.1 | 13 | 6402 | |||
XM_017025745.1 | XP_016881234.1 | 13 | 6441 | |||
XM_017025746.1 | XP_016881235.1 | 11 | 6253 | |||
XM_017025747.1 | XP_016881236.1 | 12 | 6328 | |||
XM_017025748.1 | XP_016881237.1 | 13 | 6414 | |||
XM_017025750.1 | XP_016881239.1 | 12 | 6303 |
Gene Symbol: ST8SIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005668.5 | NP_005659.1 | 5 | 3948 | ||
XM_011543630.1 | XP_011541932.1 | 4 | 3654 | |||
GenBank mRNA | BC040671.2 | - | 1 | 810 |
Gene Symbol: TLCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164407.1 | NP_001157879.1 | 4 | 4607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
