siRNA Details
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 8 | 2465 | ||
NM_198181.3 | NP_937824.3 | 9 | 2597 | |||
XM_011521601.2 | XP_011519903.1 | 8 | 3148 | |||
XM_017022226.1 | XP_016877715.1 | 7 | 2584 | |||
GenBank mRNA | AK128268.1 | - | 5 | 2144 | ||
BC041854.1 | - | 8 | 2466 | 5270411 | ||
CR936812.1 | - | 6 | 5000 |
Gene Symbol: GOLGA6L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164465.3 | NP_001157937.2 | 9 | 2918 | ||
GenBank mRNA | CR936602.1 | - | 12 | 3569 |
Gene Symbol: GOLGA6L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267536.2 | NP_001254465.2 | 9 | 3034 | ||
XM_017022481.1 | XP_016877970.1 | 10 | 2878 | |||
XM_017022482.1 | XP_016877971.1 | 10 | 2878 | |||
GenBank mRNA | AK090778.1 | - | 1 | 1139 |
Gene Symbol: GOLGA6L5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003246.4 | 10 | 2993 | |||
GenBank mRNA | BX647899.1 | - | 1 | 1390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
