siRNA Details
Gene Symbol: GOLGA6L17P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111962.1 | 6 | 627 | |||
GenBank mRNA | AK301885.1 | - | 627 |
Gene Symbol: GOLGA2P2Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001555.2 | 3 | 252 | |||
GenBank mRNA | AF332229.1 | - | 3 | 252 |
Gene Symbol: GOLGA2P3Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002195.2 | 3 | 252 |
Gene Symbol: GOLGA6L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164465.3 | NP_001157937.2 | 6 | 663 | ||
NM_001322400.1 | NP_001309329.1 | 6 | 663 | |||
GenBank mRNA | AK302092.1 | - | 6 | 625 |
Gene Symbol: GOLGA6L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310153.1 | NP_001297082.1 | 6 | 625 | ||
GenBank mRNA | AK301968.1 | - | 6 | 592 |
Gene Symbol: GOLGA6L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267536.2 | NP_001254465.2 | 6 | 625 | ||
XM_017022481.1 | XP_016877970.1 | 6 | 625 | |||
XM_017022482.1 | XP_016877971.1 | 6 | 625 |
Gene Symbol: GOLGA6L5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003246.4 | 6 | 625 | |||
GenBank mRNA | AK092490.1 | - | 3 | 238 | ||
AK097432.1 | - | 6 | 627 | |||
AK130562.1 | - | 5 | 713 | |||
DA284832.1 | - | 5 | 413 |
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 5 | 531 | ||
NM_198181.3 | NP_937824.3 | 6 | 663 | |||
XM_011521601.2 | XP_011519903.1 | 5 | 1214 | |||
GenBank mRNA | AK128268.1 | - | 2 | 212 | ||
BC041854.1 | - | 5 | 531 | 5270411 | ||
BY996849.1 | - | 3 | 406 | |||
CR936812.1 | - | 3 | 3067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
