siRNA Details
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277406.1 | NP_001264335.1 | 15 | 1918 | ||
XM_011511670.2 | XP_011509972.1 | 15 | 2090 | |||
XM_017004732.1 | XP_016860221.1 | 16 | 2069 | |||
XM_017004733.1 | XP_016860222.1 | 10 | 815 | |||
XM_017004734.1 | XP_016860223.1 | 8 | 752 |
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 15 | 1970 | ||
XM_011511216.2 | XP_011509518.1 | 11 | 1694 | |||
XM_017004159.1 | XP_016859648.1 | 11 | 1725 | |||
XM_017004160.1 | XP_016859649.1 | 7 | 693 | |||
XM_017004161.1 | XP_016859650.1 | 17 | 2407 | |||
GenBank mRNA | AY462868.1 | - | 12 | 1809 | ||
AY462871.1 | - | 15 | 1970 | |||
AY462872.1 | - | 14 | 1928 | |||
AY462873.1 | - | 13 | 1854 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 17 | 2318 | ||
XM_017004833.1 | XP_016860322.1 | 15 | 2093 | |||
XM_017004834.1 | XP_016860323.1 | 10 | 815 | |||
XM_017004835.1 | XP_016860324.1 | 8 | 752 | |||
GenBank mRNA | EF523384.1 | - | 17 | 2320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
