siRNA Details
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 1 | 336 | ||
XM_011511216.2 | XP_011509518.1 | 1 | 414 | |||
XM_017004159.1 | XP_016859648.1 | 1 | 415 | |||
XM_017004161.1 | XP_016859650.1 | 3 | 773 | |||
GenBank mRNA | AY462868.1 | - | 1 | 336 | ||
AY462871.1 | - | 1 | 336 | |||
AY462872.1 | - | 1 | 336 | |||
AY462873.1 | - | 1 | 336 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 3 | 684 | ||
XM_017004833.1 | XP_016860322.1 | 1 | 429 | |||
GenBank mRNA | EF523384.1 | - | 3 | 686 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
