siRNA Details
Gene Symbol: HLA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019111.4 | NP_061984.2 | 5 | 1263 | ||
GenBank mRNA | AK301354.1 | - | 4 | 904 | ||
BC071659.1 | - | 5 | 1202 | 5162925 | ||
J00194.1 | - | 5 | 1180 | |||
K01171.1 | - | 5 | 1168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
