siRNA Details
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 4 | 531 | ||
XM_005255741.3 | XP_005255798.1 | 5 | 1343 | |||
XM_005255742.4 | XP_005255799.1 | 5 | 1089 | |||
XM_011545929.2 | XP_011544231.1 | 6 | 1163 | |||
XM_011545931.2 | XP_011544233.1 | 5 | 796 | |||
XM_017023628.1 | XP_016879117.1 | 5 | 546 | |||
XM_017023629.1 | XP_016879118.1 | 5 | 1089 | |||
XM_017023630.1 | XP_016879119.1 | 6 | 816 | |||
XM_017023631.1 | XP_016879120.1 | 4 | 1160 | |||
XM_017023632.1 | XP_016879121.1 | 5 | 546 | |||
GenBank mRNA | BC144439.1 | - | 4 | 492 | ||
BC157836.1 | - | 4 | 1181 | 9057055 | ||
KF175222.1 | - | 4 | 493 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 4 | 439 | ||
GenBank mRNA | KF175204.1 | - | 4 | 382 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 4 | 461 | ||
XM_011523496.2 | XP_011521798.1 | 5 | 517 | |||
XM_011523498.2 | XP_011521800.1 | 5 | 551 | |||
XM_017023248.1 | XP_016878737.1 | 5 | 954 | |||
XM_017023249.1 | XP_016878738.1 | 4 | 771 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 4 | 493 | ||
XM_017023622.1 | XP_016879111.1 | 5 | 617 | |||
XM_017023623.1 | XP_016879112.1 | 5 | 617 | |||
XM_017023624.1 | XP_016879113.1 | 5 | 617 | |||
XM_017023625.1 | XP_016879114.1 | 5 | 597 | |||
XM_017023626.1 | XP_016879115.1 | 6 | 1236 | |||
XM_017023627.1 | XP_016879116.1 | 5 | 617 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 4 | 531 | ||
NM_001287251.1 | NP_001274180.1 | 5 | 617 | |||
XM_003118719.7 | XP_003118767.2 | 5 | 1092 | |||
XM_011545690.2 | XP_011543992.1 | 6 | 1181 | |||
XM_011545693.2 | XP_011543995.1 | 6 | 1180 | |||
XM_017022814.1 | XP_016878303.1 | 6 | 1181 | |||
XM_017022815.1 | XP_016878304.1 | 6 | 701 | |||
XM_017022816.1 | XP_016878305.1 | 6 | 1181 | |||
XM_017022817.1 | XP_016878306.1 | 5 | 1093 | |||
XM_017022818.1 | XP_016878307.1 | 5 | 2418 | |||
XM_017022819.1 | XP_016878308.1 | 6 | 1108 | |||
XM_017022820.1 | XP_016878309.1 | 6 | 825 | |||
XM_017022821.1 | XP_016878310.1 | 5 | 1691 | |||
XM_017022822.1 | XP_016878311.1 | 6 | 2694 | |||
XM_017022823.1 | XP_016878312.1 | 4 | 2330 | |||
XM_017022824.1 | XP_016878313.1 | 6 | 1107 | |||
XM_017022825.1 | XP_016878314.1 | 6 | 825 | |||
XM_017022826.1 | XP_016878315.1 | 5 | 642 | |||
XM_017022828.1 | XP_016878317.1 | 6 | 1180 | |||
GenBank mRNA | BC171825.1 | - | 5 | 1267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
