siRNA Details
Gene Symbol: RPS27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093614.1 | - | 1 | 1103 |
Gene Symbol: CATSPER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648099.1 | - | 9 | 2878 |
Gene Symbol: DAPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521414.1 | XP_011519716.1 | 11 | 2597 | ||
XM_017022044.1 | XP_016877533.1 | 11 | 2593 | |||
GenBank mRNA | AK094290.1 | - | 1 | 1654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
