siRNA Details
Gene Symbol: ERVK-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY371029.1 | - | 1 | 162 | ||
AY371033.1 | - | 1 | 162 | |||
AY371034.1 | - | 1 | 162 | |||
AY371035.1 | - | 1 | 162 | |||
AY371036.1 | - | 1 | 162 | |||
AY371042.1 | - | 1 | 162 | |||
AY371043.1 | - | 1 | 162 | |||
AY371044.1 | - | 1 | 162 | |||
AY371045.1 | - | 1 | 162 | |||
AY395517.1 | - | 1 | 162 | |||
AY395522.1 | - | 1 | 162 |
Gene Symbol: ERVK-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY395519.1 | - | 1 | 162 |
Gene Symbol: ERVK-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CF227268.1 | - | 2 | 375 |
Gene Symbol: ERVK-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY371031.1 | - | 1 | 162 | ||
AY371037.1 | - | 1 | 162 | |||
AY395520.1 | - | 1 | 112 | |||
AY395523.1 | - | 1 | 162 | |||
AY395524.1 | - | 1 | 162 | |||
AY395525.1 | - | 1 | 162 |
Gene Symbol: ERVK-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY371030.1 | - | 1 | 162 |
Gene Symbol: LOC105369972 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539093.2 | XP_011537395.1 | 1 | 1425 |
Gene Symbol: LOC105376906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027549.1 | XP_016883038.1 | 1 | 1059 |
Gene Symbol: LOC107986724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012900.1 | XP_016868389.1 | 1 | 1703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
