siRNA Details
Gene Symbol: MIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300867.1 | NP_001287796.1 | 23 | 2296 | ||
NM_001324062.1 | NP_001310991.1 | 29 | 5570 | |||
NM_001324063.1 | NP_001310992.1 | 26 | 5321 | |||
NM_001324064.1 | NP_001310993.1 | 29 | 5543 | |||
NM_001324065.1 | NP_001310994.1 | 24 | 2368 | |||
NM_198551.3 | NP_940953.2 | 28 | 5498 | |||
XM_006711304.3 | XP_006711367.2 | 27 | 5705 | |||
XM_017001243.1 | XP_016856732.1 | 29 | 5395 | |||
GenBank mRNA | AK025122.1 | - | 20 | 1797 | ||
AK025506.1 | - | 25 | 3178 | |||
AK026489.1 | - | 17 | 1557 | |||
AK290335.1 | - | 19 | 1773 | |||
AK296712.1 | - | 23 | 2277 | |||
AX775845.1 | - | 23 | 2270 | |||
BC047116.1 | - | 13 | 1170 | |||
D87742.1 | - | 25 | 3347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
