siRNA Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000500.7 | NP_000491.4 | 2 | 333 | ||
GenBank mRNA | AK314651.1 | - | 2 | 307 | ||
AM086564.1 | - | 2 | 223 | |||
AM086565.1 | - | 2 | 223 | |||
AM183945.1 | - | 2 | 223 | |||
AM183946.1 | - | 2 | 223 | |||
AM183947.1 | - | 2 | 223 | |||
AM183948.1 | - | 2 | 223 | |||
AM183949.1 | - | 2 | 223 | |||
AM183950.1 | - | 2 | 223 | |||
AM183951.1 | - | 2 | 223 | |||
AV708092.1 | - | 2 | 333 | |||
BC125181.1 | - | 2 | 322 | 40122017 | ||
BC125182.1 | - | 2 | 319 | 40122018 | ||
CD013985.1 | - | 1 | 381 | |||
CD013987.1 | - | 2 | 284 | |||
DA008890.1 | - | 2 | 232 | |||
DA014348.1 | - | 2 | 384 |
Gene Symbol: CYP21A1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040090.1 | 1 | 376 | |||
GenBank mRNA | CD014134.1 | - | 1 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
