siRNA Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000500.7 | NP_000491.4 | 1 | 177 | ||
NM_001128590.3 | NP_001122062.3 | 1 | 177 | |||
GenBank mRNA | AK054616.1 | - | 1 | 79 | ||
AK314651.1 | - | 1 | 151 | |||
AM086564.1 | - | 1 | 67 | |||
AM086565.1 | - | 1 | 67 | |||
AM183945.1 | - | 1 | 67 | |||
AM183946.1 | - | 1 | 67 | |||
AM183947.1 | - | 1 | 67 | |||
AM183948.1 | - | 1 | 67 | |||
AM183949.1 | - | 1 | 67 | |||
AM183950.1 | - | 1 | 67 | |||
AM183951.1 | - | 1 | 67 | |||
AV708092.1 | - | 1 | 177 | |||
BC125182.1 | - | 1 | 163 | 40122018 | ||
BP198315.1 | - | 1 | 75 | |||
CD013985.1 | - | 1 | 128 | |||
CD013987.1 | - | 1 | 128 | |||
DA008890.1 | - | 1 | 76 | |||
DA014348.1 | - | 1 | 228 |
Gene Symbol: CYP21A1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040090.1 | 1 | 123 | |||
GenBank mRNA | CD014134.1 | - | 1 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
