siRNA Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000500.7 | NP_000491.4 | 1 | 255 | ||
NM_001128590.3 | NP_001122062.3 | 1 | 255 | |||
GenBank mRNA | AK054616.1 | - | 1 | 157 | ||
CD013985.1 | - | 1 | 206 | |||
CD013987.1 | - | 1 | 206 | |||
DA008890.1 | - | 1 | 154 | |||
DA014348.1 | - | 1 | 306 |
Gene Symbol: CYP21A1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040090.1 | 1 | 201 | |||
GenBank mRNA | CD014134.1 | - | 1 | 202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
