siRNA Details
Gene Symbol: KRTAP3-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031959.2 | NP_114165.1 | 1 | 283 | ||
GenBank mRNA | AJ406932.1 | - | 1 | 283 | ||
BC034582.1 | - | 1 | 291 | 3909612 |
Gene Symbol: KRTAP3-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033185.2 | NP_149441.1 | 1 | 286 | ||
GenBank mRNA | AJ406933.1 | - | 1 | 286 | ||
BC069099.1 | - | 1 | 286 | 7216913 | ||
BC069448.1 | - | 1 | 286 | 7262684 | ||
BC093845.1 | - | 1 | 339 | 7939690 | ||
BC093847.1 | - | 1 | 339 | 7939692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
