siRNA Details
Gene Symbol: SPATA31A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015667.2 | NP_056482.2 | 4 | 1106 | ||
GenBank mRNA | BC040528.1 | - | 4 | 1106 | 4838470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
