siRNA Details
Gene Symbol: TBCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297738.1 | NP_001284667.1 | 3 | 2163 | ||
NM_001297740.1 | NP_001284669.1 | 3 | 309 | |||
NM_004607.2 | NP_004598.1 | 4 | 396 | |||
GenBank mRNA | AF038952.1 | - | 4 | 341 | ||
AK312207.1 | - | 4 | 364 | |||
BC018210.1 | - | 4 | 381 | 3861138 | ||
BC105285.1 | - | 4 | 332 | 6702573 | ||
BT007354.1 | - | 4 | 292 | |||
CR542170.1 | - | 4 | 292 | |||
CR542186.1 | - | 4 | 292 | |||
DB538817.1 | - | 2 | 159 | |||
HY015414.1 | - | 3 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
