siRNA Details
Gene Symbol: TBCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297738.1 | NP_001284667.1 | 3 | 321 | ||
NM_004607.2 | NP_004598.1 | 3 | 321 | |||
GenBank mRNA | AF038952.1 | - | 3 | 266 | ||
AK300031.1 | - | 3 | 243 | |||
AK312207.1 | - | 3 | 289 | |||
BC018210.1 | - | 3 | 306 | 3861138 | ||
BC105285.1 | - | 3 | 257 | 6702573 | ||
BT007354.1 | - | 3 | 217 | |||
CR542170.1 | - | 3 | 217 | |||
CR542186.1 | - | 3 | 217 | |||
DB538817.1 | - | 1 | 84 | |||
DC312139.1 | - | 3 | 321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
