siRNA Details
Gene Symbol: IFNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172139.2 | NP_742151.2 | 3 | 396 | ||
XM_005258765.4 | XP_005258822.1 | 4 | 808 | |||
GenBank mRNA | AY129149.1 | - | 3 | 404 | ||
AY184374.1 | - | 3 | 392 | |||
AY336714.1 | - | 3 | 395 | |||
AY336717.1 | - | 3 | 395 | |||
BC130314.1 | - | 3 | 431 | 40146345 | ||
BC130316.1 | - | 3 | 431 | 40146347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
