siRNA Details
Gene Symbol: HIST1H4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003538.3 | NP_003529.1 | 1 | 97 | ||
GenBank mRNA | BC066248.1 | - | 1 | 97 | 7002088 | |
BC066249.1 | - | 1 | 97 | 7002089 | ||
BC067495.1 | - | 1 | 97 | 7002090 | ||
BC069654.1 | - | 1 | 97 | 7262752 |
Gene Symbol: HIST1H4J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021968.3 | NP_068803.1 | 1 | 97 | ||
GenBank mRNA | AF086564.1 | - | 1 | 114 | ||
BC017361.2 | - | 1 | 116 | 4647914 | ||
BC148271.1 | - | 1 | 119 | 8860214 |
Gene Symbol: HIST1H4K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003541.2 | NP_003532.1 | 1 | 97 | ||
GenBank mRNA | AK312028.1 | - | 1 | 130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
