siRNA Details
Gene Symbol: C16orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175900.3 | NP_787096.2 | 2 | 346 | ||
GenBank mRNA | AK093000.1 | - | 2 | 333 | ||
AY358833.1 | - | 2 | 330 | |||
BC104466.2 | - | 2 | 315 | 40032034 | ||
BC104467.2 | - | 2 | 315 | 40032037 | ||
DA427069.1 | - | 2 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
